Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MTETREPAET GGYASLEEDD EDLSPGPEHS SDSEYTLSEP DSEEEEDEEE EEEETTDDPE YDPGYKVKQR LGGGRGGPSR RAPRAAQPPA QPCQLCGRSP LGEAPPGTPP CRLCCPATAP QEAPAPEGRA LGEEEEEPPR AGEGRPAGRE EEEEEEEEGT YHCTECEDSF DNLGELHGHF MLHARGEV
ZNF428 contains 1 C2H2-type zinc finger. The exact function of ZNF428 remains unknown. Two different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Enzyme-like protein PIT13; Zinc finger protein 428
Gene Aliases: C19orf37; Zfp428; ZNF428
UniProt ID: (Human) Q96B54
Entrez Gene ID: (Human) 126299
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support