Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLSDELESKP ELLVQFVQNT SIPLGQGLVE SEAKDITCLS LLPVTEASEC SRLMLPDDTT NHSNSSKEVP SSAVLRSLRV NVGPDGEETR AQTVQKSPEF LSTSESSSLL QDLQPSDSTS FILLNLTRAG LGSSAEHLVF VQDEAEDSGN DFLSSESTDS SIPWFLRVQE LAHDSLIAAT RAQLAKNAKT SSNGENVHLG SGDGQSKDSG PLPQVEKKLK CTVEGCDRTF VWPAHFKYHL KTHRNDRSFI CPAEGCGKSF YVLQRLKVHM RTHNGEKPFM CHESGCGKQF TTAGNLKNHR RIHTGEKPFL CEAQGCGRSF AEYSSLRKHL VVHSGEKPHQ CQVCGKTFSQ SGSRNVHMRK HHLQLGAAGS QEQEQTAEPL MGSSLLEEAS VPSKNLVSMN SQPSLGGESL NLPNTNSILG VDDEVLAEGS PRSLSSVPDV THHLVTMQSG RQSYEVSVLT AVNPQELLNQ GDLTERRT
Transcription factor that activates transcription of matrix-remodeling genes such as MMP1 during fibroblast senescence.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Another partner for ARF 1; clones 23667 and 23775 zinc finger protein; Zinc finger protein 410; zinc finger protein APA-1
Gene Aliases: APA-1; APA1; ZNF410
UniProt ID: (Human) Q86VK4
Entrez Gene ID: (Human) 57862
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support