Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAEAAPAPTS EWDSECLTSL QPLPLPTPPA ANEAHLQTAA ISLWTVVAAV QAIERKVEIH SRRLLHLEGR TGTAEKKLAS CEKTVTELGN QLEGKWAVLG TLLQEYGLLQ RRLENLENLL RNRNFWILRL PPGIKGDIPK VPVAFDDVSI YFSTPEWEKL EEWQKELYKN IMKGNYESLI SMDYAINQPD VLSQIQPEGE HNTEDQAGPE ESEIPTDPSE EPGISTSDIL SWIKQEEEPQ VGAPPESKES DVYKSTYADE ELVIKAEGLA RSSLCPEVPV PFSSPPAAAK DAFSDVAFKS QQSTSMTPFG RPATDLPEAS EGQVTFTQLG SYPLPPPVGE QVFSCHHCGK NLSQDMLLTH QCSHATEHPL PCAQCPKHFT PQADLSSTSQ DHASETPPTC PHCARTFTHP SRLTYHLRVH NSTERPFPCP DCPKRFADQA RLTSHRRAHA SERPFRCAQC GRSFSLKISL LLHQRGHAQE RPFSCPQCGI DFNGHSALIR HQMIHTGERP YPCTDCSKSF MRKEHLLNHR RLHTGERPFS CPHCGKSFIR KHHLMKHQRI HTGERPYPCS YCGRSFRYKQ TLKDHLRSGH NGGCGGDSDP SGQPPNPPGP LITGLETSGL GVNTEGLETN QWYGEGSGGG VL
Function as a transcriptional activator.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: zinc finger DNA binding protein p52/p71; zinc finger DNA binding protein ZER6; Zinc finger DNA-binding protein p52/p71; Zinc finger protein 398; zinc finger-estrogen receptor interaction, clone 6
Gene Aliases: KIAA1339; P51; P71; ZER6; ZNF398
UniProt ID: (Human) Q8TD17
Entrez Gene ID: (Human) 57541
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support