Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MALAPWGPVK VKKEEEEEEN FPGQASSQQV HSENIKVWAP VQGLQTGLDG SEEEEKGQNI SWDMAVVLKA TQEAPAASTL GSYSLPGTLA KSEILETHGT MNFLGAETKN LQLLVPKTEI CEEAEKPLII SERIQKADPQ GPELGEACEK GNMLKRQRIK REKKDFRQVI VNDCHLPESF KEEENQKCKK SGGKYSLNSG AVKNPKTQLG QKPFTCSVCG KGFSQSANLV VHQRIHTGEK PFECHECGKA FIQSANLVVH QRIHTGQKPY VCSKCGKAFT QSSNLTVHQK IHSLEKTFKC NECEKAFSYS SQLARHQKVH ITEKCYECNE CGKTFTRSSN LIVHQRIHTG EKPFACNDCG KAFAQSANLI VHQRSHTGEK PYECKECGKA FSCFSHLIVH QRIHTAEKPY DCSECGKAFS QLSCLIVHQR IHSGDLPYVC NECGKAFTCS SYLLIHQRIH NGEKPYTCNE CGKAFRQRSS LTVHQRTHTG EKPYECEKCG AAFISNSHLM RHHRTHLVE
ZNF35 is a new candidate transcription factor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 35; zinc finger protein 35 (clone HF.10); Zinc finger protein HF.10
Gene Aliases: HF.10; HF10; Zfp105; ZNF35
UniProt ID: (Human) P13682
Entrez Gene ID: (Human) 7584
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support