Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide Sequence: EKKESLGAAQ EALSIQLQPK ETQPFPKSEQ VYLHFLSVVT EDGPEPKDKG SLPQPPITEV ESQVFSEKLA TDTSTFEATS EGTLELQQRN PKAERLRWSP
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kruppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. ZNF232, also known as Zinc finger and SCAN domain-containing protein 11, is a 417 amino acid protein belonging to the Kruppel C2H2-type zinc-finger protein family. Localized to the nucleus, ZNF232 contains one SCAN box domain and five C2H2-type zinc fingers. Due to the presence of these domains, ZNF232 may be involved in transcriptional regulation. Ubiquitously expressed, ZNF232 is present at high levels in testis, liver and ovary. ZNF232 exists as two isoforms produced by alternative splicing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger and SCAN domain-containing protein 11; Zinc finger protein 232
Gene Aliases: ZNF232; ZSCAN11
UniProt ID: (Human) Q9UNY5
Entrez Gene ID: (Human) 7775
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support