Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide Sequence: HQEWSFQQIW EKIASDLTRS QDLVINSSQF SKEGDFPCQT EAGLSVIHTR QKSSQGNGYK PSFSDVSHFD FHQQLHSGEK SHTCDECGKN FCYISA
Matrix-assisted laser desorption ionization-time of flight analysis and examination of protein profiles from the SwissProt database revealed that the previously defined p97 repressor is ZNF224, a zinc finger protein. ZNF224, a Kruppel-like zinc finger transcription factor, is the repressor protein that specifically binds to the negative cis-element AldA-NRE and affects the AldA-NRE-mediated transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BMZF-2; Bone marrow zinc finger 2; Zinc finger protein 224; Zinc finger protein 233; Zinc finger protein 255; Zinc finger protein 27; Zinc finger protein KOX22
Gene Aliases: BMZF-2; BMZF2; KOX22; ZNF224; ZNF233; ZNF255; ZNF27
UniProt ID: (Human) Q9NZL3
Entrez Gene ID: (Human) 7767
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support