Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MRLAKPKAG ISRSSSQGKA YENKRKTGRQ RQKWGMTIRF DSSFSRLRRS LDDKPYKCTE CEKSFSQSST LFQHQKIHTG KKSHKCADCG KSFFQSSNLI QHRRIHTGEK PYKCDECGES FKQSSNLIQH QRIHTGEKPY QCDECGRCFS QSSHLIQHQR THTGEKPYQC SECGKCFSQS SHLRQHMKVH KEEKPRKTRG KNIRVKTHLP SWKAGTGRKS VAGLR (1-224 aa encoded by BC010642)
ZNF22 encodes a protein that is expressed in the epithelial component of the developing tooth organ during early bud and cap stages as well as in osteoblasts of craniofacial bone and the developing tongue.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: krox-26 protein; Zinc finger protein 22; zinc finger protein 22 (KOX 15); Zinc finger protein KOX15; Zinc finger protein Krox-26
Gene Aliases: HKR-T1; KOX15; KROX26; Zfp422; ZNF22; ZNF422
UniProt ID: (Human) P17026
Entrez Gene ID: (Human) 7570
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support