Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: PEKVHSSSEE SHACPRLKRQ LHLHQNGVEM LMENEGPQSG TNKPRETCQG PECPGLHTFL LWSHSGFNCL LCAEMLGRKE DLLHHWKHQH NCEDPSKLWA I
Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 52%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 438; ZNF438
Gene Aliases: bA330O11.1; ZNF438
UniProt ID: (Human) Q7Z4V0
Entrez Gene ID: (Human) 220929
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support