Sequence of this protein is as follows: TAACSQGNLS NCGCDREKQG YYNQAEGWKW GGCSADVRYG IDFSRRFVDA REIKKNARRL MNLHNNEAGR KVLEDRMQLE CKCHGVSGSC TTKTCWTTLP KFREVGHLLK EKYNAAVQVE VVRASRLRQP TFLRIKQLRS YQKPMETDLV YIEKSPNYCE EDAATGSVGT QGRLCNRTSP GADGCDTMCC GRGYNTHQYT KVWQCNCKFH WCCFVKCNTC SERTEVFTCK
Wnt7b encodes a member of the WNT gene family, which consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. Among members of the human WNT family, this gene product is most similar to WNT7A protein.