Immunogen sequence: EESLYLWIDA HQARVLIGFE EDILIVSEGK MAPFTHDFRK AQQRMPAIPV NIHSMNFTWQ AAGQAEYFYE FLSLRSLDKG IMADPTVNVP LLGTVPHKAS VVQVGFPCLG KQDGVAAFEV DVIVMNSEGN TILQTPQNAI
The protein encoded by this gene functions to inhibit WNT proteins, which are extracellular signaling molecules that play a role in embryonic development. This protein contains a WNT inhibitory factor (WIF) domain and five epidermal growth factor (EGF)-like domains, and is thought to be involved in mesoderm segmentation.