Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: IRESFFRVVPQ FVVFYLIYTW GTEEFERSKR KNPAAY
Highest antigen sequence identity to the following orthologs - mouse 70%, rat 68%.
This gene encodes a ubiquinone-binding protein of low molecular mass. This protein is a small core-associated protein and a subunit of ubiquinol-cytochrome c reductase complex III, which is part of the mitochondrial respiratory chain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Complex III subunit 8; Complex III subunit VIII; Cytochrome b-c1 complex subunit 8; low molecular mass ubiquinone-binding protein (9.5kD); Ubiquinol-cytochrome c reductase complex 9.5 kDa protein; Ubiquinol-cytochrome c reductase complex ubiquinone-binding protein QP-C; ubiquinol-cytochrome c reductase, complex III subunit VII, 9.5kDa
Gene Aliases: MC3DN4; QCR8; QP-C; QPC; UQCR7; UQCRQ
UniProt ID: (Human) O14949
Entrez Gene ID: (Human) 27089
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support