Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • UCP1/UCP3 Antibodies

          Invitrogen

          UCP1/UCP3 Recombinant Rabbit Monoclonal Antibody (0S10O10)

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite UCP1/UCP3 Recombinant Rabbit Monoclonal Antibody (0S10O10)

          • Antibody Testing Data (4)
          UCP1/UCP3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          UCP1/UCP3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          UCP1/UCP3 Antibody (MA5-37913) in ICC/IF

          Immunofluorescence analysis of UCP1/UCP3 in rat brown adipose cells. Samples were incubated with UCP1/UCP3 Monoclonal antibody (Product # MA5-37913) using a dilution of 1:100 (40x lens). Blue: DAPI for nuclear staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          UCP1/UCP3 Antibody in Immunocytochemistry (ICC/IF)
          UCP1/UCP3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UCP1/UCP3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UCP1/UCP3 Antibody in Western Blot (WB)

          Product Details

          MA5-37913

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          0S10O10

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 1-307 of mouse UCP1+UCP3 (P12242)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol, 0.05% BSA

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2897833

          Product Specific Information

          Positive Samples: Mouse skeletal muscle, Mouse fat

          Immunogen sequence: MVNPTTSEVQ PTMGVKIFSA GVSACLADII TFPLDTAKVR LQIQGEGQAS STIRYKGVLG TITTLAKTEG LPKLYSGLPA GIQRQISFAS LRIGLYDSVQ EYFSSGRETP ASLGNKISAG LMTGGVAVFI GQPTEVVKVR MQAQSHLHGI KPRYTGTYNA YRVIATTESL STLWKGTTPN LMRNVIINCT ELVTYDLMKG ALVNNKILAD DVPCHLLSAL VAGFCTTLLA SPVDVVKTRF INSLPGQYPS VPSCAMSMYT KEGPTAFFKG FVASFLRLGS WNVIMFVCFE QLKKELMKSR QTVDCTT

          Target Information

          UCP are mitochondrial transporter proteins that create proton leaks across the inner mitochondrial membrane, thus uncoupling oxidative phosphorylation. As a result, energy is dissipated in the form of heat. May play a role in the modulation of tissue respiratory control. Participates in thermogenesis and energy balance.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Mitochondrial brown fat uncoupling protein 1; mitochondrial uncoupling protein 3; Putative mitochondrial transporter UCP3; Solute carrier family 25 member 7; Solute carrier family 25 member 9; Thermogenin; UCP 1; UCP 3; uncoupling protein 1, mitochondrial; Uncoupling protein 3; Uncoupling protein 3, mitochondrial; uncoupling protein, mitochondrial; Uncoupling protein-3

          View more View less

          Gene Aliases: AI385626; AI645527; Slc25a7; Slc25a9; Ucp; UCP-3; Ucp1; Ucp3; Ucpa; Uncp

          View more View less

          UniProt ID: (Rat) P04633, (Mouse) P12242, (Mouse) P56501, (Rat) P56499

          View more View less

          Entrez Gene ID: (Rat) 24860, (Mouse) 22227, (Mouse) 22229, (Rat) 25708

          View more View less

          Function(s)
          oxidative phosphorylation uncoupler activity protein binding hydrogen ion transmembrane transporter activity
          Process(es)
          response to superoxide response to hypoxia fatty acid metabolic process transport mitochondrial transport proton transport transmembrane transport generation of precursor metabolites and energy regulation of transcription from RNA polymerase II promoter cellular response to hormone stimulus positive regulation of multicellular organismal metabolic process response to steroid hormone aging response to nutrient response to cold response to activity response to insulin response to glucocorticoid hydrogen ion transmembrane transport
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-66966984d8-2vwff:80/100.66.134.75:80.
          git-commit: 4f7892df6eac4c3aaa23131f1084e28be0ddeb54
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0