Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MATEAPVNIA PPECSTVVST AVDSLIWQPN SLNMHMIRPK SAKGRTRPSL QKSQGVEVCA HHIPSPPPAI PYELPSSQKP GACAPKSPNQ GASDEIPELQ QQVPTGASSS LNKYPVLPSI NRKNLEEEAV ETVAKKASSL QLSSIRALYQ DETGTMKTSE EDSRARACAV ERKFIVRTKK QGSSRAGNLE EPSDQEPRLL LAVRSPTGQR FVRHFRPTDD LQTIVAVAEQ KNKTSYRHCS IETMEVPRRR FSDLTKSLQE CRIPHKSVLG ISLEDGEGWP
UBXN10 (VCP/p97) binding protein required for ciliogenesis. Acts as a tethering factor that facilitates recruitment of VCP/p97 to the intraflagellar transport complex B (IFT-B) in cilia. UBX domain-containing proteins act as tethering factors for VCP/p97 and may specify substrate specificity of VCP/p97.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: UBX domain containing 3; UBX domain-containing protein 10; UBX domain-containing protein 3
Gene Aliases: UBXD3; UBXN10
UniProt ID: (Human) Q96LJ8
Entrez Gene ID: (Human) 127733
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support