Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MQESQETHIS NHLDEVVAAV SITHRKKFQN KLLQTALFQP PREKLHLCEE KAKSYSNSHE YKQAVHELVR CVALTRICYG DSHWKLAEAH VNLAQGYLQL KGLSLQAKQH AEKARQILAN SIVPPYSENT DVFKFSIELF HTMGRALLSL QKFKEAAENL TKAERLSKEL LQCGRIIKEE WIEIEARIRL SFAQVYQGQK KSKEALSHYQ AALEYVEISK GETSRECVPI LRELAGVEQA LGLHDVSINH FLQAHLIILS RSPSQVEAAD SAHIVAHAAV ASGRHEHHDV AEQYFQESMA HLKDSEGMGR TKFLSIQDEF CHFLQMTGQK ERATSILRES LEAKVEAFGD FSPEVAETYR LLGGADLAQG NHSGARKKLK KGDLSFLLSA TLP
Participates positively in the ciliary Hedgehog (Hh) signaling. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cervical cancer proto-oncogene 8 protein; HCC-8; Tetratricopeptide repeat protein 23; TPR repeat protein 23
Gene Aliases: HCC-8; HCC8; TTC23
UniProt ID: (Human) Q5W5X9
Entrez Gene ID: (Human) 64927
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support