Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEDAGGGEET PAPEAPHPPQ LAPPEEQGLL FQEETIDLGG DEFGSEENET ASEGSSPLAD KLNEHMMESV LISDSPNSEG DAGDLGRVRD EAEPGGEGDP GPEPAGTPSP SGEADGDCAP EDAAPSSGGA PRQDAAREVP GSEAARPEQE PPVAEPVPVC TIFSQRAPPA SGDGFEPQMV KSPSFGGASE ASARTPPQVV QPSPSLSTFF GDTAASHSLA SDFFDSFTTS AFISVSNPGA GSPAPASPPP LAVPGTEGRP EPVAMRGPQA AAPPASPEPF AHIQAVFAGS DDPFATALSM SEMDRRNDAW LPGEATRGVL RAVATQQRGA VFVDKENLTM PGLRFDNIQG DAVKDLMLRF LGEKAAAKRQ VLNADSVEQS FVGLKQLISC RNWRAAVDLC GRLLTAHGQG YGKSGLLTSH TTDSLQLWFV RLALLVKLGL FQNAEMEFEP FGNLDQPDLY YEYYPHVYPG RRGSMVPFSM RILHAELQQY LGNPQESLDR LHKVKTVCSK ILANLEQGLA EDGGMSSVTQ EGRQASIRLW RSRLGRVMYS MANCLLLMKD YVLAVDAYHS VIKYYPEQEP QLLSGIGRIS LQIGDIKTAE KYFQDVEKVT QKLDGLQGKI MVLMNSAFLH LGQNNFAEAH RFFTEILRMD PRNAVANNNA AVCLLYLGKL KDSLRQLEAM VQQDPRHYLH ESVLFNLTTM YELESSRSMQ KKQALLEAVA GKEGDSFNTQ CLKLA
Component of the TRAPP complex, which is involved in endoplasmic reticulum to Golgi apparatus trafficking at a very early stage (PubMed:21525244, PubMed:28777934). Also plays a role in chromosome congression, kinetochore assembly and stability and controls the recruitment of CENPE to the kinetochores (PubMed:25918224). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: tetratricopeptide repeat domain 15; Tetratricopeptide repeat protein 15; TPR repeat protein 15; Trafficking of membranes and mitosis; Trafficking protein particle complex subunit 12
Gene Aliases: CGI-87; TRAMM; TRAPPC12; TTC-15; TTC15
UniProt ID: (Human) Q8WVT3
Entrez Gene ID: (Human) 51112
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support