Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: KDHKSHNVSL IEEAAQNYQG QIQEQIQVLQ QKEKETVQVK AQGVHRVDVF TDQVEHEKQR ILTEFELLHQ VLEEEKNFLL SRIYWLGHEG TEAGKHYVAS TEPQLNDLKK LVDSLKTKQN MPPRQLLEDI KVVLCRSEEF QFLNPTPVPL ELEKKLSEAK SRHDSITGSL KKFKDQLQAD RKKDENRFFK SMNKNDMKSW GLLQKNNHKM NKTSEPGSSS AGGRTTSGPP NHHSSAPSHS LFRASSAGKV TFPVCLLASY DEISGQGASS
TRIM31 encodes for a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to both the cytoplasm and the nucleus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase TRIM31; tripartite motif-containing 31; Tripartite motif-containing protein 31
Gene Aliases: C6orf13; HCG1; HCGI; RNF; TRIM31
UniProt ID: (Human) Q9BZY9
Entrez Gene ID: (Human) 11074
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support