Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MTDQQAEAR SYLSEEMIAE FKAAFDMFDA DGGGDISVKE LGTVMRMLGQ TPTKEELDAI IEEVDEDGSG TIDFEEFLVM MVRQMKEDAK GKSEEELAEC FRIFDRNADG YIDPEELAEI FRASGEHVTD EEIESLMKDG DKNNDGRIDF DEFLKMMEGV Q (1-160 aa encoded by BC005323)
Troponin (Tn), a key protein complex in the regulation of striated muscle contraction, is composed of 3 subunits. The Tn-I subunit inhibits actomyosin ATPase, the Tn-T subunit binds tropomyosin and Tn-C, while the Tn-C subunit binds calcium and overcomes the inhibitory action of the troponin complex on actin filaments. The protein encoded by this gene is the Tn-C subunit.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: skeletal troponin C; STNC; troponin C type 2 (fast); troponin C, fast skeletal; Troponin C, skeletal muscle; troponin C2, fast
Gene Aliases: Tncs; TNNC2
UniProt ID: (Human) P02585, (Mouse) P20801
Entrez Gene ID: (Human) 7125, (Rat) 296369, (Mouse) 21925
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support