Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSLMLDDQPP MEAQYAEEGP GPGIFRAEPG DQQHPISQAV CWRSMRRGCA VLGALGLLAG AGVGSWLLVL YLCPAASQPI SGTLQDEEIT LSCSEASAEE ALLPALPKTV SFRINSEDFL LEAQVRDQPR WLLVCHEGWS PALGLQICWS LGHLRLTHHK GVNLTDIKLN SSQEFAQLSP RLGGFLEEAW QPRNNCTSGQ VVSLRCSECG ARPLASRIVG GQSVAPGRWP WQASVALGFR HTCGGSVLAP RWVVTAAHCM HSFRLARLSS WRVHAGLVSH SAVRPHQGAL VERIIPHPLY SAQNHDYDVA LLRLQTALNF SDTVGAVCLP AKEQHFPKGS RCWVSGWGHT HPSHTYSSDM LQDTVVPLFS TQLCNSSCVY SGALTPRMLC AGYLDGRADA CQGDSGGPLV CPDGDTWRLV GVVSWGRGCA EPNHPGVYAK VAEFLDWIHD TAQDSLL
This gene encodes a protein that belongs to the serine protease family. Serine proteases are known to be involved in many physiological and pathological processes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC141886; MGC148044; Spinesin; Transmembrane protease serine 5
Gene Aliases: SPINESIN; TMPRSS5
UniProt ID: (Human) Q9H3S3
Entrez Gene ID: (Human) 80975
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support