Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: PPPLPSALPAM PSVPTVSSLG GLNFPWMESS RRFVKDRFTA
Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
The Hox proteins play a role in patterns of embryonic development and cellular differentiation by regulating downstream target genes. TLX3, also known as homeobox11-like1 (HOX11L2), is a DNA-binding nuclear transcription factor that is expressed in a subset of the primary sensory nervous system. TLX3, along with the related TLX1, is a selector transcription factor that promotes an excitatory glutamatergic neuronal phenotype over an inhibitory GABAergic phenotype opposing LBX1 signals during dorsal spinal cord development. Chromosomal translocations of the TLX3 gene have been shown to result in some forms of T-cell acute lymphoblastic leukemia (T-ALL).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support