Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: MATRSSDWLR RPLGEDNQPE TQLFWDKEPW FWHDTLTEQL WRIFAGVHDE KAKPRDRQQA PGLGQESKAP GSCDPGTDPC PEDASTPRPP EASSSPPEGS QDRNTSWGVV QEPPGRASRF LQSISWDPED FEDAWKRPDA LPGQSKRLAV PCKLEKMRIL AHGELVLATA ISSFTRHVFT CGRRGIKVWS LTGQVAEDRF PESHLPIQTP GAFLRTCLLS SNSRSLLTGG YNLASVSVWD LAAPSLHVKE QLPCAGLNCQ ALDANLDANL AFASFTSGVV RIWDLRDQSV VRDLKGYPDG VKSIVVKGYN IWTGGPDACL RCWDQRTIMK PLEYQFKSQI MSLSHSPQED WVLLGMANGQ QWLQSTSGSQ RHMVGQKDSV ILSVKFSPFG QWWASVGMDD FLGVYSMPAG TKVFEVPEMS PVTCCDVSSN NRLVVTGSGE HASVYQITY
As a member of the subcortical maternal complex (SCMC), plays an essential role for zygotes to progress beyond the first embryonic cell divisions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: transducin-like enhancer of split 6 (E(sp1) homolog, Drosophila); Transducin-like enhancer protein 6
Gene Aliases: GRG6; PREMBL; TLE6
UniProt ID: (Human) Q9H808
Entrez Gene ID: (Human) 79816
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support