Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • TIRAP Antibodies

          Invitrogen

          TIRAP Recombinant Rabbit Monoclonal Antibody (9T1A8)

          View all (20) TIRAP antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite TIRAP Recombinant Rabbit Monoclonal Antibody (9T1A8)

          • Antibody Testing Data (3)
          TIRAP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          TIRAP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TIRAP Antibody (MA5-38196) in IHC (P)

          Immunohistochemistry analysis of TIRAP in paraffin-embedded mouse kidney. Samples were incubated with TIRAP Monoclonal antibody (Product # MA5-38196) using a dilution of 1:100 (40x lens). Perform microwave antigen retrieval with 10 mM Tris/EDTA buffer pH 9.0 before commencing with IHC staining protocol. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TIRAP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TIRAP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TIRAP Antibody in Western Blot (WB)

          Product Details

          MA5-38196

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          9T1A8

          Immunogen

          A synthetic peptide corresponding to a sequence within amino acids 1-100 of human TIRAP (P58753)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.02 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 0.05% BSA, 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2898113

          Product Specific Information

          Positive Samples: 293T, HepG2, THP-1, Mouse brain, Rat spleen, Rat thymus

          Immunogen sequence: MASSTSLPAP GSRPKKPLGK MADWFRQTLL KKPKKRPNSP ESTSSDASQP TSQDSPLPPS LSSVTSPSLP PTHASDSGSS RWSKDYDVCV CHSEEDLVAA

          Target Information

          Toll-like receptors (TLRs) are signaling molecules that recognize different microbial products during infection and serve as an important link between the innate and adaptive immune responses. These proteins act through adaptor molecules such as TIRAP and MyD88 to activate various kinases and transcription factors. In TIRAP-deficient mice, TLR signaling in response to TLR2 ligands (using either TLR1 and TLR6 as co-receptors) is totally abolished, suggesting that MyD88 and TIRAP work together and are both required for TLR2 signaling. Furthermore, these mice are also resistant to the toxic effects of LPS and show defects in NF-βB and MAP kinase activation, suggesting that TIRAP is also involed in TLR4 signaling.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: adapter protein wyatt; Adaptor protein Wyatt; FLJ42305; HGNC:17192; MyD88 adapter-like protein; MyD88-2; OTTHUMP00000179130; TIR domain-containing adapter protein; toll-interleukin 1 receptor (TIR) domain containing adaptor protein; toll-interleukin 1 receptor (TIR) domain-containing adaptor protein; Toll-like receptor 4 adaptor protein; Toll-like receptor adaptor protein; Toll/interleukin-1 receptor domain-containing adapter protein

          View more View less

          Gene Aliases: AA407980; BACTS1; C130027E04Rik; MAL; MyD88-2; TIRAP; Tlr4ap; Wyatt

          View more View less

          UniProt ID: (Human) P58753, (Mouse) Q99JY1

          View more View less

          Entrez Gene ID: (Human) 114609, (Mouse) 117149, (Rat) 680127

          View more View less

          Function(s)
          protein kinase C binding protein binding phosphatidylinositol-4,5-bisphosphate binding protein binding, bridging Toll-like receptor 4 binding Toll-like receptor 2 binding protein homodimerization activity protein heterodimerization activity molecular_function
          Process(es)
          immune system process inflammatory response signal transduction cell surface receptor signaling pathway I-kappaB kinase/NF-kappaB signaling activation of JUN kinase activity response to bacterium myeloid cell differentiation positive regulation of B cell proliferation response to lipopolysaccharide regulation of interferon-beta production regulation of interleukin-15 production positive regulation of interleukin-12 production positive regulation of interleukin-15 production positive regulation of interleukin-8 production positive regulation of tumor necrosis factor production positive regulation of toll-like receptor 2 signaling pathway positive regulation of toll-like receptor 3 signaling pathway positive regulation of toll-like receptor 4 signaling pathway toll-like receptor 5 signaling pathway TIRAP-dependent toll-like receptor 4 signaling pathway positive regulation of I-kappaB kinase/NF-kappaB signaling negative regulation of growth of symbiont in host innate immune response regulation of innate immune response positive regulation of innate immune response positive regulation of interleukin-6 biosynthetic process positive regulation of JNK cascade defense response to Gram-positive bacterium positive regulation of NF-kappaB transcription factor activity positive regulation of ERK1 and ERK2 cascade interleukin-1-mediated signaling pathway 3'-UTR-mediated mRNA stabilization cellular response to bacterial lipopeptide cellular response to lipoteichoic acid positive regulation of neutrophil chemotaxis positive regulation of protein homodimerization activity positive regulation of chemokine (C-X-C motif) ligand 1 production positive regulation of chemokine (C-X-C motif) ligand 2 production MyD88-dependent toll-like receptor signaling pathway activation of NF-kappaB-inducing kinase activity toll-like receptor 4 signaling pathway biological_process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-66966984d8-2vwff:80/100.66.134.75:80.
          git-commit: 4f7892df6eac4c3aaa23131f1084e28be0ddeb54
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0