PA1-803 detects endogenous Tpit from mouse AtT-20 cell lysates.
PA1-803 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~48 kDa protein representing Tpit from AtT-20 cells.
PA1-803 immunizing protein, bacterially produced, corresponds to the mouse C-terminal region of Tpit.
This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. Mutations in this gene were found in patients with isolated deficiency of pituitary POMC-derived ACTH, suggesting an essential role for this gene in differentiation of the pituitary POMC lineage. ACTH deficiency is characterized by adrenal insufficiency symptoms such as weight loss, lack of appetite (anorexia), weakness, nausea, vomiting, and low blood pressure.