Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Cas9 Antibodies

          GenScript

          SpCas9 Monoclonal Antibody (14B6), GenCRISPR™

          View all (49) Cas9 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite SpCas9 Monoclonal Antibody (14B6), GenCRISPR™

          • Antibody Testing Data (8)
          SpCas9 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SpCas9 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 8

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SpCas9 Antibody (A01936-40) in ICC/IF

          Immunofluorescence staining of GenCRISPR™ SpCas9 Antibody (14B6) (GenScript, A01936-40) with SpCas9 in Hela cells. Hela cells transfected with PX458 (pSpCas9(BB)-2A-GFP) (green) were fixed with 4% Poly-Formaldehyde (5min) and then blocked in 3% BSA 30min. The cells were then incubated with GenCRISPR™ SpCas9 Antibody (14B6) (GenScript, A01936-40) at 2 µg/mL, 1 µg/mL and mouse IgG at 2 µg/mL, 1 µg/mL at room temperature for 2h, followed by a further incubation at 37 °C for 1h with Goat Anti-Mouse IgG Antibody (H&L) [ifluor 555], pAb (GenScri... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SpCas9 Antibody in Immunocytochemistry (ICC/IF)
          SpCas9 Antibody in Western Blot (WB)
          SpCas9 Antibody in Western Blot (WB)
          SpCas9 Antibody in Western Blot (WB)
          SpCas9 Antibody in Western Blot (WB)
          SpCas9 Antibody in Western Blot (WB)
          SpCas9 Antibody in ELISA (ELISA)
          SpCas9 Antibody in ELISA (ELISA)

          Product Details

          A01936-40

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.5-2 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          1-2 µg/mL
          -

          ELISA (ELISA)

          0.05-1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Bacteria

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          14B6

          Immunogen

          Recombinant Streptococcus pyogenes CRISPR/Cas9

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C or -80°C if preferred

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitute the lyophilized antibody with deionized water (or equivalent) to a final concentration of 0.5 mg/ml.

          Clustered regularly interspaced short palindromic repeats (CRISPR)/CRISPR-associated (Cas) protein 9 system provides a robust and multiplexable genome editing tool, enabling researchers to precisely manipulate specific genomic elements, and facilitating the elucidation of target gene function in biology and diseases. CRISPR/Cas9 genome editing allows for double-stranded DNA breaks at specific sequences to efficiently disrupt, excise, mutate, insert, or replace genes. To date, the Streptococcus pyogenes Cas9 (SpCas9) has been used broadly to achieve efficient genome editing in a variety of species and cell types. It is important that the precision of transfection and the level of Cas9 expression should be controlled strictly during the editing processes by using specific anti-CRISPR/Cas9 antibodies.

          Target Information

          Cas9 is an RNA-guided DNA endonuclease enzyme associated with the CRISPR (Clustered Regularly Interspaced Short Palindromic Repeats) adaptive immunity system in bacteria and genetic engineering. The system is unique and flexible due to its dependence on RNA as the element that targets the nuclease to a desired DNA sequence. It can be used to induce indel mutations, specific sequence replacements/insertions, and large deletions or genomic rearrangements at any desired location in the genome. In addition, Cas9 can also be used to mediate upregulation of specific endogenous genes or to alter histone modifications or DNA methylation. Cas9 may be derived from Streptococcus pyogenes (SpCas9) or Staphylococcus aureus (SaCas9). SaCas9 has a number of properties that make it advantageous for genome editing, including its small size, high efficiency, nickase activity, and apparent specificity. SaCas9 principally recognizes an NNGRRT protospacer adjacent motif (PAM) and cleaves target DNA at rates comparable to SpCas9 with an ideal spacer lengths of 24 to 20 nucleotides. In comparison to SpCas9, SaCas9 is smaller (1368 AA versus 1053 AA) and generates indels at a comparable rate when directed against target DNA with the mutually permissive NGGRRT PAM. This makes it the preferred enzyme for all-in-one delivery of Cas9 and multiple gRNA expression cassettes with Adeno-associated vectors.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
          git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0