Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Probable transcription factor required during spermatogenesis and oogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: helix-loop-helix protein NOHLH; newborn ovary helix loop helix; RP11-100C15.3; spermatogenesis associated 27; Spermatogenesis- and oogenesis-specific basic helix-loop-helix-containing protein 1
Gene Aliases: bA100C15.3; bHLHe80; C9orf157; Gm110; NOHLH; RGD1564440; SOHLH1; SPATA27; TEB2; Tohlh1
UniProt ID: (Human) Q5JUK2, (Mouse) Q6IUP1
Entrez Gene ID: (Human) 402381, (Mouse) 227631, (Rat) 362085
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support