Search Thermo Fisher Scientific
Biologo
Positive control: Gastric and intestinal epithelia.
Reactivity: Human and other primates (no reactivity in chicken, cat, rabbit, rat, sheep and goat).
Serotonin (also known as 5-hydroxytryptamne or 5-HT) is a biochemical messenger and neurotransmitter synthesized from L-tryptophan. Serotonin has a range of functions, including regulation of gastric motility, and may be related to some psychiatric disorders including schizophrenia and depression. Serotoinin is widely distributed in the central and peripheral nervous system, and also in some non-nueral tissues such as platelets.