Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • SUSD2 Antibodies

          Abnova

          SUSD2 Polyclonal Antibody, MaxPab™

          View all (5) SUSD2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite SUSD2 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          SUSD2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SUSD2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SUSD2 Antibody (H00056241-B01P) in WB

          Western Blot analysis of SUSD2 expression in transfected 293T cell line by SUSD2 MaxPab polyclonal antibody. Lane 1: SUSD2 transfected lysate(90.42 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SUSD2 Antibody in Western Blot (WB)

          Product Details

          H00056241-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SUSD2 (NP_062547.1, 1 a.a. approximately 822 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MKPALLPWAL LLLATALGPG PGPTADAQES CSMRCGALDG PCSCHPTCSG LGTCCLDFRD FCLEILPYSG SMMGGKDFVV RHFKMSSPTD ASVICRFKDS IQTLGHVDSS GQVHCVSPLL YESGRIPFTV SLDNGHSFPR AGTWLAVHPN KVSMMEKSEL VNETRWQYYG TANTSGNLSL TWHVKSLPTQ TITIELWGYE ETGMPYSQEW TAKWSYLYPL ATHIPNSGSF TFTPKPAPPS YQRWRVGALR IIDSKNYAGQ KDVQALWTND HALAWHLSDD FREDPVAWAR TQCQAWEELE DQLPNFLEEL PDCPCTLTQA RADSGRFFTD YGCDMEQGSV CTYHPGAVHC VRSVQASLRY GSGQQCCYTA DGTQLLTADS SGGSTPDRGH DWGAPPFRTP PRVPSMSHWL YDVLSFYYCC LWAPDCPRYM QRRPSNDCRN YRPPRLASAF GDPHFVTFDG TNFTFNGRGE YVLLEAALTD LRVQARAQPG TMSNGTETRG TGLTAVAVQE GNSDVVEVRL ANRTGGLEVL LNQEVLSFTE QSWMDLKGMF LSVAAGDRVS IMLASGAGLE VSVQGPFLSV SVLLPEKFLT HTHGLLGTLN NDPTDDFTLH SGRVLPPGTS PQELFLFGAN WTVHNASSLL TYDSWFLVHN FLYQPKHDPT FEPLFPSETT LNPSLAQEAA KLCGDDHFCN FDVAATGSLS TGTATRVAHQ LHQRRMQSLQ PVVSCGWLAP PPNGQKEGNR YLAGSTIYFH CDNGYSLAGA ETSTCQADGT WSSPTPKCQP GRSYAVLLGI IFGGLAVVAA VALVYVLLRR RKGNTHVWGA QP

          Target Information

          May be a cytokine receptor for C10ORF99. May be a tumor suppressor; together with C10ORF99 has a growth inhibitory effect on colon cancer cells which includes G1 cell cycle arrest (PubMed:25351403). May play a role in breast tumorigenesis (PubMed:23131994).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Sushi domain (SCR repeat) containing; Sushi domain-containing protein 2; testicular tissue protein Li 190

          View more View less

          Gene Aliases: BK65A6.2; SUSD2

          View more View less

          UniProt ID: (Human) Q9UGT4

          View more View less

          Entrez Gene ID: (Human) 56241

          View more View less

          Function(s)
          scavenger receptor activity protein binding polysaccharide binding
          Process(es)
          receptor-mediated endocytosis immune response negative regulation of cell division negative regulation of cell cycle G1/S phase transition
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
          git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0