Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MSSQQQKQP CIPPPQLQQQ QVKQPCQPPP QEPCIPKTKE PCHPKVPEPC HPKVPEPCQP KVPEPCHPKV PEPCPSIVTP APAQQKTKQK (1-89 aa encoded by BC056240 )
SPRR1B is a keratinocyte protein that first appears in the cell cytosol, but ultimately becomes cross-linked to membrane proteins by transglutaminase. All that results in the formation of an insoluble envelope beneath the plasma membrane. Can function as both amine donor and acceptor in transglutaminase-mediated cross-linkage.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 14.9 kDa pancornulin; cornifin B; Cornifin-B; small proline-rich protein 1B; Small proline-rich protein IB; SPR-IB
Gene Aliases: CORNIFIN; GADD33; SPR-IB; SPRR1; SPRR1B
UniProt ID: (Human) P22528
Entrez Gene ID: (Human) 6699
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support