Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MAFAPMGPE ASFFDVLDRH RESLLAALRR GGREPPTGGS RLASRFEDSV GLQMVSHCTT RKIKSDSPKS AQKFSLILKI LSMIYKLVQS NTYATKRDIY YTDSQLFGNQ TVVDNIINDI SCMLKVSRRS LHILSTSKGL IAGNLRYIEE DGTKVNCTCG ATAVAVPSNI QGIRNLVTDA KFVLIVEKDA TFQRLLDDNF CNKLSPCIMI TGKGVPDLNT RLLVKKLWDT FHVPVFTLVD ADPHGIEIMC IYKYGSMSMS FEAHHLTVPA IRWLGLLPSD LKRLNVPKDS LIPLTKRDQM KLDSILRRPY VTCQPFWRKE MEIMADSKMK AEIQALTFLS SDYLSRVYLP NKLKFGGWI (1-358 aa encoded by BC033591 )
Meiotic recombination and chromosome segregation require the formation of double-strand breaks (DSBs) in paired chromosome homologs. During meiosis in yeast, a meiotic recombination protein is covalently-linked to the 5' end of DSBs and is essential for the formation of DSBs. The protein encoded by this gene is similar in sequence and conserved features to the yeast meiotic recombination protein. The encoded protein belongs to the TOP6A protein family. Several transcript variants encoding different isoforms have been found for this gene, but the full-length nature of only two of them have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 35; CT35; Meiotic recombination protein SPO11; MGC39953; spermatogenesis associated 43; SPO11 meiotic protein covalently bound to DSB homolog; sporulation protein, meiosis-specific, SPO11 homolog (S. cervisiae)
Gene Aliases: AI449549; CT35; SPATA43; SPO11; TOPVIA
UniProt ID: (Human) Q9Y5K1, (Mouse) Q9WTK8
Entrez Gene ID: (Human) 23626, (Mouse) 26972
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support