Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MKLSGMFLLL SLALFCFLTG VFSQGGQVDC GEFQDTKVYC TRESNPHCGS DGQTYGNKCA FCKAIVKSGG KISLKHPGKC
SPINK6 (serine peptidase inhibitor, Kazal type 6), also known as BUSI2, is an 80 amino acid secreted protein that contains one kazal-like domain and is thought to function as a serine protease inhibitor. The gene encoding SPINK6 maps to human chromosome 5, which contains 181 million base pairs and comprises nearly 6% of the human genome. Chromosome 5 is associated with Cockayne syndrome through the ERCC8 gene and familial adenomatous polyposis through the adenomatous polyposis coli (APC) tumor suppressor gene. Treacher Collins syndrome is also associated with chromosome 5 and is caused by insertions or deletions within the TCOF1 gene. Deletion of the p arm of chromosome 5 leads to Cri du chat syndrome, while deletion of the q arm or of chromosome 5 altogether is common in therapy-related acute mye-logenous leukemias and myelodysplastic syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acrosin inhibitor; Kallikrein inhibitor; protease inhibitor H; Serine protease inhibitor Kazal-type 6
Gene Aliases: BUSI2; SPINK6; UNQ844; UNQ844/PRO1782
UniProt ID: (Human) Q6UWN8
Entrez Gene ID: (Human) 404203
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support