Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The function of this protein has not been specifically defined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adapter-related protein complex 5 zeta subunit; adaptor related protein complex 5, zeta 1 subunit; Adaptor-related protein complex 5 zeta subunit; adaptor-related protein complex 5, zeta 1 subunit; AP-5 complex subunit zeta-1; protein KIAA0415; Zeta5
Gene Aliases: AP5Z1; KIAA0415; SPG48; zeta
UniProt ID: (Human) O43299
Entrez Gene ID: (Human) 9907
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support