Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • SPERT Antibodies

          Proteintech

          SPERT Polyclonal Antibody

          View all (6) SPERT antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite SPERT Polyclonal Antibody

          • Antibody Testing Data (6)
          SPERT Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SPERT Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SPERT Antibody (17826-1-AP) in ICC/IF

          Immunofluorescent analysis of (4% PFA) fixed PC-3 cells using SPERT antibody (Product # 17826-1-AP) at dilution of 1:200 and CoraLite®488-Conjugated AffiniPure Goat Anti-Rabbit IgG(H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SPERT Antibody in Immunocytochemistry (ICC/IF)
          SPERT Antibody in Immunocytochemistry (ICC/IF)
          SPERT Antibody in Western Blot (WB)
          SPERT Antibody in Western Blot (WB)
          SPERT Antibody in Western Blot (WB)
          SPERT Antibody in Western Blot (WB)

          Product Details

          17826-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          SPERT Fusion Protein Ag12200 (1-412 aa encoded by BC027609)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.29 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MQPEGLKCR MEESNAERGT AEPFPRLHNL YSTPRCAQQA ALPRLSRRMA SQHSYPLNRF SSVPLDPMER PMSQADLELD YNPPRVQLSD EMFVFQDGRW VNENCRLQSP YFSPSASFHH KLHHKRLAKE CMLQEENKSL REENKALREE NRMLSKENKI LQVFWEEHKA SLGREESRAP SPLLHKDSAS LEVVKKDHVA LQVPRGKEDS TLQLLREENR ALQQLLEQKQ AYWAQAEDTA APAEESKPAP SPHEEPCSPG LLQDQGSGLS SRFEEPKGPP ARQEDSKELR ALRKMVSNMS GPSGEEEAKV GPGLPDGCQP LQLLREMRQA LQALLKENRL LQEENRTLQV LRAEHRGFQE ENKALWENNK LKLQQKLVID TVTEVTARME MLIEELYAFM PARSQDPKKP SRV (1-412 aa encoded by BC027609)

          Target Information

          SPERT (spermatid associated protein), also known as CBY2 (chibby homolog 2) or NURIT, is a novel leucine-zipper protein belonging to the chibby family of proteins. Uniquely Expressed in the spermatid flower-like structure, SPERT interacts with Nek1, a member of the NIMA-family kinase family that is associated centrosomal stability and ciliogenesis. It contains a leucine-zipper motif and two coiled-coil regions, and is transcribed through the elongation stage of the spermatids. SPERT is absent from mature spermatozoa and is thought to be involved in transporting proteins that are to be discarded via the residual bodies.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: chibby family member 2; chibby homolog 2; novel leucine zipper testicular protein; Protein chibby homolog 2; Protein nurit; spermatid flower-like structure protein; Spermatid-associated protein; testicular tissue protein Li 182; testis-specific leucine zipper protein nurit

          View more View less

          Gene Aliases: 1700086N05Rik; AI596460; CBY2; NURIT; SPERT

          View more View less

          UniProt ID: (Human) Q8NA61, (Mouse) Q32MG2, (Rat) Q6AXV6

          View more View less

          Entrez Gene ID: (Human) 220082, (Mouse) 67926, (Rat) 498572

          View more View less

          Function(s)
          protein binding molecular_function
          Process(es)
          biological_process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5fd7d668bc-tpg7z:80/100.66.134.75:80.
          git-commit: 4d16a52b65a0a59161c0e81e148ee2d2ac6d3307
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc