Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
SPATA21 is involved in the differentiation of haploid spermatids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Spergen-3; Spermatogenesis-associated protein 21; Spermatogenic cell-specific gene 3 protein
Gene Aliases: 4933414G08; SPATA21; spergen-2; spergen2; Spergen3
UniProt ID: (Human) Q7Z572, (Rat) Q68A65, (Mouse) Q8BHW6
Entrez Gene ID: (Human) 374955, (Rat) 366491, (Mouse) 329972
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support