PA5-31662 targets SNX27 in IHC (P) and WB applications and shows reactivity with Human and mouse samples.
The PA5-31662 immunogen is recombinant fragment corresponding to a region within amino acids 74 and 288 of Human SNX27.
SNX27 encodes a member of the sorting nexin family, a diverse group of cytoplasmic and membrane-associated proteins involved in endocytosis of plasma membrane receptors and protein trafficking through these compartments. All members of this protein family contain a phosphoinositide binding domain (PX domain). A highly similar protein in mouse is responsible for the specific recruitment of an isoform of serotonin 5-hydroxytryptamine 4 receptor into early endosomes, suggesting the analogous role for the human protein.