Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEGEPPPVEE RRRLQEELNE FVESGCRTLE EVTASLGWDL DSLDPGEEEA AEDEVVICPY DSNHHMPKSS LAKHMASCRL RKMGYTKEEE DEMYNPEFFY ENVKIPSITL NKDSQFQIIK QARTAVGKDS DCYNQRIYSS LPVEVPLNHK RFVCDLTQAD RLALYDFVVE ETKKKRSDSQ IIENDSDLFV DLAAKINQDN SRKSPKSYLE ILAEVRDYKR RRQSYRAKNV HITKKSYTEV IRDVINVHME ELSNHWQEEQ EKAEDDAEKN EERRSASVDS RQSGGSYLDA ECSRHRRDRS RSPHKRKRNK DKDKNCESRR RKERDGERHH SHKRRKQKI
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: small nuclear ribonucleoprotein 48kDa (U11/U12); small nuclear ribonucleoprotein, U11/U12 48KDa subunit; U11/U12 small nuclear ribonucleoprotein 48 kDa protein; U11/U12 snRNP 48 kDa protein; U11/U12 snRNP 48K; U11/U12-48K
Gene Aliases: C6orf151; dJ336K20B.1; dJ512B11.2; SNRNP48
UniProt ID: (Human) Q6IEG0
Entrez Gene ID: (Human) 154007
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support