Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MEGEPPPVE ERRRLQEELN EFVESGCRTL EEVTASLGWD LDSLDLGEEE AAEDEVVICP YDSNHHMPKS SLAKHMASCR LRKMGYTKEE EDEMYNPEFF YENVKIPSIT LNKDSQFQII KQARTAVGKD SDCYNQRIYS SLPVEVPLNH KRFVCDLTQA DRLALYDFVV EETKKKRSDS QIIENDSDLF VDLAAKINQD NSRKSPKSYL EILAEVRDYK RRRQSYRAKN VHITKKSYTE VIRDVINVHM EELSNHWQEE QEKAEDDAEK NEERRSASVD SRQSGGSYLD AECSRHRRDR SRSPHKRKRN KDKDKNCESR RRKERDGERH HSHKRRKQKI (1-339 aa encoded by BC114630)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: small nuclear ribonucleoprotein 48kDa (U11/U12); small nuclear ribonucleoprotein, U11/U12 48KDa subunit; U11/U12 small nuclear ribonucleoprotein 48 kDa protein; U11/U12 snRNP 48 kDa protein; U11/U12 snRNP 48K; U11/U12-48K
Gene Aliases: 1110050F08Rik; 6530403A03Rik; C6orf151; dJ336K20B.1; dJ512B11.2; SNRNP48
UniProt ID: (Human) Q6IEG0, (Mouse) Q9D361
Entrez Gene ID: (Human) 154007, (Mouse) 67797
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support