PA5-31920 targets SMYD3 in IHC (P) and WB applications and shows reactivity with Human samples.
The PA5-31920 immunogen is recombinant protein fragment corresponding to a region within amino acids 201 and 411 of Human SMYD3.
SMYD3 (histone-lysine N-methyltransferase) methylates Lys-4 of histone H3 and Lys-5 of histone H4. It induces di- and tri-methylation. SMYD3 plays an important role in transcriptional activation as a member of an RNA polymerase complex.