Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MSIEIESSDV IRLIMQYLKE NSLHRALATL QEETTVSLNT VDSIESFVAD INSGHWDTVL QAIQSLKLPD KTLIDLYEQV VLELIELREL GAARSLLRQT DPMIMLKQTQ PERYIHLENL LARSYFDPRE AYPDGSSKEK RRAAIAQALA GEVSVVPPSR LMALLGQALK WQQHQGLLPP GMTIDLFRGK AAVKDVEEEK FPTQLSRHIK FGQKSHVECA RFSPDGQYLV TGSVDGFIEV WNFTTGKIRK DLKYQAQDNF MMMDDAVLCM CFSRDTEMLA TGAQDGKIKV WKIQSGQCLR RFERAHSKGV TCLSFSKDSS QILSASFDQT IRIHGLKSGK TLKEFRGHSS FVNEATFTQD GHYIISASSD GTVKIWNMKT TECSNTFKSL GSTAGTDITV NSVILLPKNP EHFVVCNRSN TVVIMNMQGQ IVRSFSSGKR EGGDFVCCAL SPRGEWIYCV GEDFVLYCFS TVTGKLERTL TVHEKDVIGI AHHPHQNLIA TYSEDGLLKL WKP
SMU1 (suppressor of Mec8 and Unc-52 homolog), also known as BWD (brain-enriched WD), is a member of the WD-repeat SMU1 family and contains one CTLH domain, one LisH domain and six WD-repeats. SMU1 is a homolog of the C. elegans protein Smu1, a ubiquitously expressed nuclear protein that is believed to play a role in alternative splicing events of Unc-52. In mammals, SMU1 is a component of the spliceosome and appears to exhibit a conserved function, affecting the splicing of the mammalian Unc-52 homolog, namely Perlecan. Mutations in the gene encoding SMU1 may affect the function of the spliceosome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brain-enriched WD-repeat protein; functional spliceosome-associated protein 57; smu-1 suppressor of mec-8 and unc-52 homolog; Smu-1 suppressor of mec-8 and unc-52 protein homolog; WD40 repeat-containing protein SMU1
Gene Aliases: BWD; fSAP57; SMU-1; SMU1
UniProt ID: (Human) Q2TAY7
Entrez Gene ID: (Human) 55234
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support