Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: KVAKDCIRFL KEERAEPETF LALDYLDIKP INERLRELKG CKMVIDVIKT QFPQLKKVIQ FVCGNGLVCE TMEEARHIAL SGPERQKTVA LDGTLFLKSG VISGGSSDLK
The SMC1B gene encodes a meiosis-specific subunit of the cohesin complex, which is essential for proper chromosome segregation during meiosis. It plays a critical role in sister chromatid cohesion and DNA recombination. The SMC1B protein, also known as SMC1 beta, forms a heterodimer with SMC3, which is necessary for the formation of the cohesin complex that holds sister chromatids together until anaphase. Studies have demonstrated that SMC1B is required for the activation of the spindle assembly checkpoint (SAC) during oocyte meiosis. The loss of SMC1B function leads to improper chromosome alignment and segregation, resulting in infertility due to meiotic arrest. Furthermore, the gene is expressed exclusively during meiosis, and mutations or deletions within SMC1B can cause sterility due to disruptions in meiotic progression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bK268H5; bK268H5.5; mitosis-specific chromosome segregation protein like protein beta; RP1-102D24.3; SMC protein 1B; SMC-1-beta; SMC-1B; SMC1 (structural maintenance of chromosomes 1, yeast)-like 1; SMC1 structural maintenance of chromosomes 1-like 2; Structural maintenance of chromosomes protein 1B
Gene Aliases: SMC1B; SMC1BETA; SMC1L2
UniProt ID: (Human) Q8NDV3
Entrez Gene ID: (Human) 27127
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support