FIGURE: 1 / 4
Product is shipped at room temperature as a lyophilized powder and should be stored at -20C upon receipt.
Reconstitute using 50 uL deionized water.
Slc30a3 encodes a protein that is involved in accumulation of zinc in synaptic vesicles.