Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: EAAHFLFGEP TLRKRKSPAQ VMFQCLWKSC GKVLSTASAM QRHIRLVHLG RQAEPEQSDG EEDFYYTELD VGVDTLTDGL SSLTPVSPTA SM
Transcription factor involved in SLC2A4 and HD gene transactivation. Binds to the consensus sequence 5'-GCCGGCG-3'.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GEF; GLUT4 enhancer factor; HDBP-1; Huntington disease gene regulatory region-binding protein 1; Huntington's disease gene regulatory region-binding protein 1; SLC2A4 regulator
Gene Aliases: GEF; HDBP-1; HDBP1; Si-1-2; Si-1-2-19; SLC2A4RG
UniProt ID: (Human) Q9NR83
Entrez Gene ID: (Human) 56731
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support