Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MQSHIGWQNM PSLWASAQDV WNTRGRKLLL IYRGGSLVIL RSSVTWGLTT AIHDFLQRKS HSRKELKTD
Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 30%.
SLC25A53 (Solute Carrier Family 25 Member 53) is a Protein Coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial carrier triple repeat protein 6; Solute carrier family 25 member 53; solute carrier family 25, member 53
Gene Aliases: MCART6; SLC25A53
UniProt ID: (Human) Q5H9E4
Entrez Gene ID: (Human) 401612
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support