Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MPVPASWPHL PSPFLLMTLL LGRLTGVAGE DELQVIQPEK SVSVAAGESA TLRCAMTSLI PVGPIMWFRG AGAGRELIYN QKEGHFPRVT TVSELTKRNN LDFSISISNI TPADAGTYYC VKFRKGSPDD VEFKSGAGTE LSVREPALAP TAPLLVALLL GPKLLLVVGV SAIYICWKQK A
SIRP beta 1 is a type I transmembrane protein belonging to the SIRP family within the Ig superfamily. Members of this family are characterized by an extracellular region containing a V-set Ig domain with a J-like sequence and two C1-set Ig domains. Unlike SIRP alpha that has cytoplasmic ITIM domains, SIRP beta 1 possesses positively charged residues that allow association with ITAM motif-containing adaptor molecules. SIRP beta 1 is expressed on cells of monocyte, macrophage or dendritic lineages.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD172 antigen-like family member B; CD172b; CD172b antigen; DKFZp686A05192; signal-regulatory protein beta 1; Signal-regulatory protein beta-1; Signal-regulatory protein beta-1 isoform 3; SIRP BETA 1; SIRP beta 1 cell surface protein; SIRP-beta B; SIRP-beta-1; SIRP-beta-1 isoform 3
Gene Aliases: 9930027N05Rik; CD172b; SIRP-beta; SIRP-BETA-1; Sirpb; SIRPB1
UniProt ID: (Human) O00241
Entrez Gene ID: (Human) 10326, (Mouse) 320832
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support