Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: RTVVMRPSVS WEKAGPEEAK APVRGDEAPP ARVAGPAAGT PPCQMGVYPT DLTLQLLAVR RKSRLRDPGL QQTLR
Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 63%.
SH3TC1 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SH3 domain and tetratricopeptide repeat-containing protein 1; SH3 domain and tetratricopeptide repeats-containing protein 1
Gene Aliases: SH3TC1
UniProt ID: (Human) Q8TE82
Entrez Gene ID: (Human) 54436
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support