Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MAMQAAKRA NIRLPPEVNR ILYIRNLPYK ITAEEMYDIF GKYGPIRQIR VGNTPETRGT AYVVYEDIFD AKNACDHLSG FNVCNRYLVV LYYNANRAFQ KMDTKKKEEQ LKLLKEKYGI NTDPPK (1-125 aa encoded by B C015463)
SF3B14 encodes a 14 kDa protein subunit of the splicing factor 3b complex. Splicing factor 3b associates with both the U2 and U11/U12 small nuclear ribonucleoprotein complexes (U2 snRNP) of spliceosomes. This 14 kDa protein interacts directly with subunit 1 of the splicing factor 3b complex. This 14 kDa protein also interacts directly with the adenosine that carries out the first transesterification step of splicing at the pre-mRNA branch site.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: pre mRNA branch site protein p14; Pre-mRNA branch site protein p14; SF3B 14 kDa subunit; SF3B14a; spliceosome-associated protein, 14 kDa subunit; Spliceosome-associated protein, 14-kDa; Splicing factor 3B subunit 6; splicing factor 3B, 14 kDa subunit; splicing factor 3B, 14 kDa subunit-like; Splicing factor 3b, subunit 6, 14kDa
Gene Aliases: 0610009D07Rik; 6030419K15Rik; AV001342; CGI-110; HSPC175; HT006; P14; SAP14; SAP14a; SF3B14; SF3B14A; SF3B6
UniProt ID: (Human) Q9Y3B4, (Mouse) P59708
Entrez Gene ID: (Human) 51639, (Mouse) 66055, (Rat) 100365504
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support