Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MQTCPLAFPG HVSQALGTLL FLAASLSAQN EGWDSPICTE GVVSVSWGEN TVMSCNISNA FSHVNIKLRA HGQESAIFNE VAPGYFSRDG WQLQVQGGVA QLVIKGARDS HAGLYMWHLV GHQRNNRQVT LEVSGAEPQS APDTGFWPVP AVVTAVFILL VALVMFAWYR CRCSQQRREK KFFLLEPQMK VAALRAGAQQ GLSRASAELW TPDSEPTPRP LALVFKPSPL GALELLSPQP LFPYAADP
This gene encodes a transmembrane and secreted protein with characteristics of a type 1a transmembrane protein. It is found in a perinuclear Golgi-like pattern and thought to be involved in hematopoietic and/or immune system processes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein K-12; Secreted and transmembrane protein 1; type 1a transmembrane protein
Gene Aliases: K12; SECTM1
UniProt ID: (Human) Q8WVN6
Entrez Gene ID: (Human) 6398
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support