PA1-25702 detects SAR1 from human, mouse, and rat samples.
PA1-25702 has been successfully used in Western blot procedures.
The PA1-25702 immunogen is residues 1-198 of SAR1.
Involved in transport from the endoplasmic reticulum to the Golgi apparatus. Required to maintain SEC16A localization at discrete locations on the ER membrane perhaps by preventing its dissociation. SAR1A-GTP-dependent assembly of SEC16A on the ER membrane forms an organized scaffold defining endoplasmic reticulum exit sites (ERES). [UniProt]