Antibody tested in indirect ELISA
Rhodamine B is a fluorescent dye molecule that belongs to the rhodamine family. It is commonly used as a tracer molecule in various fields such as biology, chemistry, and materials science. Rhodamine B has a high quantum yield, meaning it emits strong fluorescence signals, and it is also relatively stable and easy to handle. Its bright and intense color, along with its excellent photostability and chemical stability, makes it an excellent choice for use in fluorescence microscopy, flow cytometry, and other imaging techniques.