PA5-34827 targets Renin in ICC, IF, and WB applications and shows reactivity with Human samples.
The PA5-34827 immunogen is recombinant fragment corresponding to a region within amino acids 1 and 302 of Renin (Uniprot ID#P00797).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Renin is a highly specific endopeptidase, whose only known function is to generate angiotensin I from angiotensinogen in the plasma, initiating a cascade of reactions that produce an elevation of blood pressure and increased sodium retention by the kidney.