Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • RUNX2 Antibodies

          Abnova

          RUNX2 Polyclonal Antibody, MaxPab™

          View all (42) RUNX2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite RUNX2 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          RUNX2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          RUNX2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RUNX2 Antibody (H00000860-D01P) in WB

          Western Blot analysis of RUNX2 expression in transfected 293T cell line by RUNX2 MaxPab polyclonal antibody. Lane 1: RUNX2 transfected lysate(55.77 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RUNX2 Antibody in Western Blot (WB)

          Product Details

          H00000860-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          RUNX2 (AAI60022.1, 1 a.a. approximately 507 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MRIPVDPSTS RRFSPPSSSL QPGKMSDVSP VVAAQQQQQQ QQQQQQQQQQ QQQQQQQEAA AAAAAAAAAA AAAAAVPRLR PPHDNRTMVE IIADHPAELV RTDSPNFLCS VLPSHWRCNK TLPVAFKVVA LGEVPDGTVV TVMAGNDENY SAELRNASAV MKNQVARFND LRFVGRSGRG KSFTLTITVF TNPPQVATYH RAIKVTVDGP REPRRHRQKL DDSKPSLFSD RLSDLGRIPH PSMRVGVPPQ NPRPSLNSAP SPFNPQGQSQ ITDPRQAQSS PPWSYDQSYP SYLSQMTSPS IHSTTPLSST RGTGLPAITD VPRRISDDDT ATSDFCLWPS TLSKKSQAGA SELGPFSDPR QFPSISSLTE SRFSNPRMHY PATFTYTPPV TSGMSLGMSA TTHYHTYLPP PYPGSSQSQS GPFQTSSTPY LYYGTSSGSY QFPMVPGGDR SPSRMLPPCT TTSNGSTLLN PNLPNQNDGV DADGSHSSSP TVLNSSGRMD ESVWRPY

          Target Information

          RUNX2, a nuclear protein, is a member of the RUNX family of transcription factors and has an Runt DNA-binding domain. It is essential for membranous and endochondral bone formation. It not only regulates osteoblastic differentiation and skeletal morphogenesis but also acts as a scaffold for nucleic acids and regulatory factors involved in skeletal gene expression. The protein can bind DNA both as a monomer and with more affinity, as a subunit of a heterodimeric complex. It plays a critical role in increasing TGFBR1 expression by osteoblasts and functions in cooperation with DLX5 or a related factor to activate osteoblast-specific gene expression. Mutations lead to disorders in bone development like cleidocranial dysplasia (CCD).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Acute myeloid leukemia 3 protein; AML-3; CBF-alpha-1; Core-binding factor subunit alpha-1; core-binding factor, runt domain, alpha subunit 1; Oncogene AML-3; OSF-2; Osteoblast-specific transcription factor 2; PEA2-alpha A; PEBP2-alpha A; Polyomavirus enhancer-binding protein 2 alpha A subunit; Runt-related transcription factor 2; RUNX 2; RUNX-2; SL3-3 enhancer factor 1 alpha A subunit; SL3/AKV core-binding factor alpha A subunit

          View more View less

          Gene Aliases: AML3; CBF-alpha-1; CBFA1; CCD; CCD1; CLCD; OSF-2; OSF2; PEA2aA; PEBP2A; PEBP2aA; RUNX2

          View more View less

          UniProt ID: (Human) Q13950

          View more View less

          Entrez Gene ID: (Human) 860

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II core promoter sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding chromatin binding transcription factor activity, sequence-specific DNA binding protein binding ATP binding protein domain specific binding histone deacetylase binding bHLH transcription factor binding repressing transcription factor binding
          Process(es)
          ossification osteoblast differentiation endochondral ossification osteoblast fate commitment chondrocyte differentiation chondrocyte development osteoblast development regulation of transcription from RNA polymerase II promoter transcription from RNA polymerase II promoter positive regulation of cell proliferation hemopoiesis neuron differentiation T cell differentiation BMP signaling pathway positive regulation of chondrocyte differentiation embryonic forelimb morphogenesis regulation of fibroblast growth factor receptor signaling pathway odontogenesis of dentin-containing tooth regulation of odontogenesis of dentin-containing tooth cellular response to fibroblast growth factor stimulus regulation of cell differentiation positive regulation of osteoblast differentiation negative regulation of smoothened signaling pathway negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated cell maturation embryonic cranial skeleton morphogenesis stem cell differentiation cellular response to BMP stimulus positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-7p7bh:80/100.66.135.138:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0