Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAYSTVQRVA LASGLVLALS LLLPKAFLSR GKRQEPPPTP EGKLGRFPPM MHHHQAHSDG QTPGARFQRS HLAEAFAKAK GSGGGAGGGG SGRGLMGQII PIYGFGIFLY ILYILFKLSK GKTTAEDGKC YTAMPGNTHR KITSFELAQL QEKLKETEAA MEKLFNRVGP NGERAQTVTS DQEKRLLHQL REITRVMKEG KFIDRFSPEK EAEEAPYMED WEGYPEETYP IYDLSDCIKR RQETILVDYP DPKELSAEEI AERMGMIEEE ESDHLGWESL PTDPRAQEDN SVTSCDPKPE TCSCCFHEDE DPAVLAENAG FSADSYPEQE ETTKEEWSQD FKDEGLGIST DKAYTGSMLR KRNPQGLE
Promotes functional expression of homomeric alpha-7 and alpha-8 nicotinic acetylcholine receptors at the cell surface. May also promote functional expression of homomeric serotoninergic 5- HT3 receptors, and of heteromeric acetylcholine receptors alpha- 3/beta-2, alpha-3/beta-4, alpha-4/beta-2 and alpha-4/beta-4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein RIC-3; resistance to inhibitors of cholinesterase 3 homolog; resistance to inhibitors of cholinesterase 3-like protein; Resistant to inhibitor of cholinesterase 3
Gene Aliases: AYST720; PRO1385; RIC3; UNQ720/PRO1385
UniProt ID: (Human) Q7Z5B4
Entrez Gene ID: (Human) 79608
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support